Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAX1
Entrez Gene ID
506895
Gene Name
HCLS1 associated protein X-1
Gene Aliases
HAX-1
Chromosome Location
Chr.3: 16250187 - 16253025 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045953.2 NP_001039418.2 6 - 7 853 115
GenBank mRNA BC112670.1 - 6 - 7 809 115
EV706642.1 - 1 - 2 280 115

View Full Product Details