Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTR1
Entrez Gene ID
513180
Gene Name
endosome associated trafficking regulator 1
Gene Aliases
SDCCAG3
Chromosome Location
Chr.11: 103858807 - 103865120 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046140.2 NP_001039605.1 1 - 2 219 52
GenBank mRNA BC105395.1 - 1 - 2 220 52

View Full Product Details