Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C13H20orf27
Entrez Gene ID
515589
Gene Name
chromosome 13 C20orf27 homolog
Gene Aliases
-
Chromosome Location
Chr.13: 51430704 - 51443748 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046205.2 NP_001039670.1 5 - 6 636 73
GenBank mRNA BC112587.1 - 5 - 6 637 73

View Full Product Details