Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIMM9
Entrez Gene ID
521073
Gene Name
translocase of inner mitochondrial membrane 9
Gene Aliases
-
Chromosome Location
Chr.10: 70683244 - 70699660 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046254.1 NP_001039719.1 2 - 3 318 119
GenBank mRNA BC112498.1 - 2 - 3 318 119

View Full Product Details