Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DKKL1
Entrez Gene ID
540124
Gene Name
dickkopf like acrosomal protein 1
Gene Aliases
-
Chromosome Location
Chr.18: 55842565 - 55847428 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046469.2 NP_001039934.1 4 - 5 581 87
GenBank mRNA BC110180.1 - 4 - 5 582 87

View Full Product Details