Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHL5
Entrez Gene ID
615499
Gene Name
four and a half LIM domains 5
Gene Aliases
-
Chromosome Location
Chr.9: 53181874 - 53239359 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076379.2 NP_001069847.1 2 - 3 313 94
GenBank mRNA BC110187.1 - 2 - 3 314 94

View Full Product Details