Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHCHD6
Entrez Gene ID
615934
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 6
Gene Aliases
-
Chromosome Location
Chr.22: 60288808 - 60388647 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076402.2 NP_001069870.1 6 - 7 629 90
GenBank mRNA BC109788.1 - 6 - 7 630 90

View Full Product Details