Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNDC8
Entrez Gene ID
617117
Gene Name
fibronectin type III domain containing 8
Gene Aliases
-
Chromosome Location
Chr.19: 14964939 - 14976767 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076444.2 NP_001069912.1 2 - 3 647 89
GenBank mRNA BC110216.1 - 2 - 3 648 89

View Full Product Details