Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC58
Entrez Gene ID
508149
Gene Name
coiled-coil domain containing 58
Gene Aliases
-
Chromosome Location
Chr.1: 66798342 - 66819186 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083389.2 NP_001076858.1 2 - 3 178 125
GenBank mRNA BC114911.1 - 2 - 3 179 125

View Full Product Details