Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPA1
Entrez Gene ID
505093
Gene Name
heterogeneous nuclear ribonucleoprotein A1
Gene Aliases
HNRPA1, hnRNP A1
Chromosome Location
Chr.5: 25779263 - 25785721 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045911.1 NP_001039376.1 4 - 5 552 74
GenBank mRNA BC105413.1 - 4 - 5 552 74

View Full Product Details