Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAPTM5
Entrez Gene ID
512361
Gene Name
lysosomal protein transmembrane 5
Gene Aliases
HA1520
Chromosome Location
Chr.2: 122807359 - 122833480 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046118.2 NP_001039583.1 1 - 2 152 68
GenBank mRNA BC105438.1 - 1 - 2 154 68

View Full Product Details