Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMG1
Entrez Gene ID
515362
Gene Name
EMG1 N1-specific pseudouridine methyltransferase
Gene Aliases
-
Chromosome Location
Chr.5: 103487265 - 103492985 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046197.2 NP_001039662.2 4 - 5 599 74
GenBank mRNA BC113268.1 - 2 - 3 1017 74
DV843519.1 - 4 - 5 600 74
EV745816.1 - 2 - 3 443 74

View Full Product Details