Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PON1
Entrez Gene ID
523798
Gene Name
paraoxonase 1
Gene Aliases
-
Chromosome Location
Chr.4: 12542349 - 12576241 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046269.2 NP_001039734.1 3 - 4 211 123
GenBank mRNA BC112488.1 - 3 - 4 213 123
EU289337.1 - 3 - 4 123

View Full Product Details