Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C29H11orf54
Entrez Gene ID
530784
Gene Name
chromosome 29 C11orf54 homolog
Gene Aliases
C11orf54
Chromosome Location
Chr.29: 1056362 - 1080757 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046315.2 NP_001039780.1 6 - 7 546 88
GenBank mRNA BC105390.1 - 6 - 7 547 88

View Full Product Details