Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LSM5
Entrez Gene ID
615136
Gene Name
LSM5 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
-
Chromosome Location
Chr.4: 64014906 - 64018570 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046553.2 NP_001040018.1 3 - 4 195 145
GenBank mRNA BC105404.1 - 3 - 4 196 145

View Full Product Details