Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
N4BP2L1
Entrez Gene ID
616069
Gene Name
NEDD4 binding protein 2 like 1
Gene Aliases
-
Chromosome Location
Chr.12: 28591515 - 28614285 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046574.1 NP_001040039.1 2 - 3 338 91
GenBank mRNA BC113273.1 - 2 - 3 338 91

View Full Product Details