Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD17B13
Entrez Gene ID
618192
Gene Name
hydroxysteroid 17-beta dehydrogenase 13
Gene Aliases
-
Chromosome Location
Chr.6: 102192495 - 102212283 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046616.2 NP_001040081.1 4 - 5 666 70
GenBank mRNA BC114051.1 - 4 - 5 667 70

View Full Product Details