Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAS2R42
Entrez Gene ID
664646
Gene Name
taste receptor, type 2, member 42
Gene Aliases
T2R67
Chromosome Location
Chr.5: 98558817 - 98559755 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046633.1 NP_001040098.1 1 - 1 650 81

View Full Product Details