Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
T2R65A
Entrez Gene ID
664647
Gene Name
bitter taste receptor Bota-T2R65A
Gene Aliases
-
Chromosome Location
Chr.5: 98625559 - 98626446 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046634.1 NP_001040099.1 1 - 1 747 107

View Full Product Details