Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNASE2
Entrez Gene ID
282218
Gene Name
deoxyribonuclease 2, lysosomal
Gene Aliases
-
Chromosome Location
Chr.7: 12676596 - 12679150 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075127.2 NP_001068595.1 5 - 6 853 112
GenBank mRNA AF047017.1 - 3 - 4 448 112
BC118429.1 - 5 - 6 854 112

View Full Product Details