Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIST1
Entrez Gene ID
506589
Gene Name
intestine-specific transcript 1 protein
Gene Aliases
-
Chromosome Location
Chr.7: 4940162 - 4984503 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075271.1 NP_001068739.1 3 - 4 718 99

View Full Product Details