Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIRT4
Entrez Gene ID
519328
Gene Name
sirtuin 4
Gene Aliases
-
Chromosome Location
Chr.17: 62622997 - 62639895 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075785.1 NP_001069253.1 1 - 2 31 87
GenBank mRNA BC116055.1 - 1 - 2 31 87

View Full Product Details