Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FH
Entrez Gene ID
520260
Gene Name
fumarate hydratase
Gene Aliases
-
Chromosome Location
Chr.16: 35260924 - 35288031 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075803.2 NP_001069271.1 3 - 4 400 93
GenBank mRNA BC118450.1 - 3 - 4 401 93

View Full Product Details