The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075804.2 | NP_001069272.1 | 1 - 2 | 157 | 105 |
GenBank mRNA | BC114881.1 | - | 1 - 2 | 158 | 105 |