Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPM1
Entrez Gene ID
534097
Gene Name
dolichyl-phosphate mannosyltransferase subunit 1, catalytic
Gene Aliases
DPMS
Chromosome Location
Chr.13: 78852506 - 78869898 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076013.2 NP_001069481.1 1 - 2 176 74
GenBank mRNA BC116151.1 - 1 - 2 177 74
GQ367549.1 - 1 - 2 165 74

View Full Product Details