Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPT4
Entrez Gene ID
617915
Gene Name
angiopoietin 4
Gene Aliases
ANGPTL4
Chromosome Location
Chr.13: 60191143 - 60234886 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076483.2 NP_001069951.1 4 - 5 1370 89
GenBank mRNA BC114033.1 - 4 - 5 1372 89
EU599227.1 - 4 - 5 1110 89

View Full Product Details