Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAP1L5
Entrez Gene ID
508508
Gene Name
nucleosome assembly protein 1 like 5
Gene Aliases
-
Chromosome Location
Chr.6: 36082759 - 36084691 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077878.2 NP_001071346.1 1 - 1 1844 128
GenBank mRNA BC114819.1 - 1 - 1 1845 128
BC142424.1 - 1 - 1 2052 128

View Full Product Details