Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VIM
Entrez Gene ID
280955
Gene Name
vimentin
Gene Aliases
-
Chromosome Location
Chr.13: 31653816 - 31661745 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173969.3 NP_776394.2 6 - 7 1351 78
GenBank mRNA BC118269.1 - 6 - 7 1351 78
L13263.1 - 6 - 7 1223 78

View Full Product Details