The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075328.1 | NP_001068796.1 | 9 - 10 | 1534 | 115 |
GenBank mRNA | BC123444.1 | - | 9 - 10 | 1534 | 115 |
HM347499.1 | - | 9 - 10 | 1459 | 115 |