Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRSN1
Entrez Gene ID
510831
Gene Name
neurensin 1
Gene Aliases
-
Chromosome Location
Chr.23: 33501362 - 33521675 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075469.1 NP_001068937.1 3 - 4 376 111
GenBank mRNA BC119900.1 - 3 - 4 376 111

View Full Product Details