Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTCRA
Entrez Gene ID
513669
Gene Name
pre T cell antigen receptor alpha
Gene Aliases
-
Chromosome Location
Chr.23: 16547283 - 16552639 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075633.1 NP_001069101.1 1 - 2 205 97
GenBank mRNA BC120034.1 - 1 - 2 205 97

View Full Product Details