Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCTN6
Entrez Gene ID
513751
Gene Name
dynactin subunit 6
Gene Aliases
-
Chromosome Location
Chr.27: 26320287 - 26342213 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075638.2 NP_001069106.1 3 - 4 215 82
GenBank mRNA BC118351.1 - 3 - 4 216 82

View Full Product Details