Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GABRA5
Entrez Gene ID
523515
Gene Name
gamma-aminobutyric acid type A receptor subunit alpha5
Gene Aliases
-
Chromosome Location
Chr.21: 4267289 - 4359565 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075844.1 NP_001069312.1 4 - 5 556 108
GenBank mRNA BC123418.1 - 4 - 5 556 108

View Full Product Details