Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAAF1
Entrez Gene ID
533348
Gene Name
proteasomal ATPase associated factor 1
Gene Aliases
WDR71
Chromosome Location
Chr.15: 53344605 - 53386028 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075990.1 NP_001069458.1 10 - 11 1034 110
GenBank mRNA BC118307.1 - 10 - 11 1034 110
BT025484.1 - 10 - 11 1017 110

View Full Product Details