Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMM1
Entrez Gene ID
537070
Gene Name
phosphomannomutase 1
Gene Aliases
-
Chromosome Location
Chr.5: 112654716 - 112665041 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076083.1 NP_001069551.1 1 - 2 442 95
GenBank mRNA BC120394.1 - 1 - 2 442 95

View Full Product Details