The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076113.2 | NP_001069581.1 | 5 - 6 | 923 | 65 |
GenBank mRNA | BC120203.1 | - | 5 - 6 | 924 | 65 |