Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC30A3
Entrez Gene ID
512803
Gene Name
solute carrier family 30 member 3
Gene Aliases
-
Chromosome Location
Chr.11: 72369362 - 72382875 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076843.1 NP_001070311.1 4 - 5 714 102
GenBank mRNA BC123455.1 - 4 - 5 714 102

View Full Product Details