Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SARAF
Entrez Gene ID
515461
Gene Name
store-operated calcium entry associated regulatory factor
Gene Aliases
TMEM66
Chromosome Location
Chr.27: 26248900 - 26273960 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076856.1 NP_001070324.1 5 - 6 1118 99
GenBank mRNA BC123456.1 - 5 - 6 1118 99

View Full Product Details