Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB31
Entrez Gene ID
616979
Gene Name
RAB31, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.24: 41786924 - 41868539 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076944.1 NP_001070412.1 4 - 5 358 69
GenBank mRNA BC123831.1 - 4 - 5 358 69

View Full Product Details