Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNRC2
Entrez Gene ID
767836
Gene Name
proline rich nuclear receptor coactivator 2
Gene Aliases
-
Chromosome Location
Chr.2: 128938565 - 128942609 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076957.1 NP_001070425.1 1 - 2 156 65
GenBank mRNA BC119962.1 - 1 - 2 156 65

View Full Product Details