Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RESP18
Entrez Gene ID
510410
Gene Name
regulated endocrine specific protein 18
Gene Aliases
-
Chromosome Location
Chr.2: 107270818 - 107276358 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077897.2 NP_001071365.1 2 - 3 269 64
GenBank mRNA BC126772.1 - 2 - 3 271 64

View Full Product Details