Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AQP3
Entrez Gene ID
780866
Gene Name
aquaporin 3
Gene Aliases
-
Chromosome Location
Chr.8: 75194697 - 75200652 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079794.1 NP_001073262.1 4 - 5 532 67
GenBank mRNA BC123791.1 - 4 - 5 550 67

View Full Product Details