Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX4
Entrez Gene ID
519597
Gene Name
sorting nexin 4
Gene Aliases
-
Chromosome Location
Chr.1: 69893690 - 69946619 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080281.2 NP_001073750.1 7 - 8 749 94
GenBank mRNA BC126650.1 - 7 - 8 750 94

View Full Product Details