Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOD3
Entrez Gene ID
532481
Gene Name
superoxide dismutase 3
Gene Aliases
EC-SOD, ECSOD
Chromosome Location
Chr.6: 44372154 - 44393167 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082610.1 NP_001076079.1 2 - 3 1011 72
GenBank mRNA AY957404.1 - 2 - 3 1011 72

View Full Product Details