Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKAPD1
Entrez Gene ID
513716
Gene Name
NKAP domain containing 1
Gene Aliases
C15H11orf57
Chromosome Location
Chr.15: 22382967 - 22395429 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083423.1 NP_001076892.1 5 - 6 972 150
GenBank mRNA BC123788.1 - 5 - 6 972 150

View Full Product Details