The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083424.1 | NP_001076893.1 | 4 - 5 | 345 | 97 |
GenBank mRNA | BC123685.1 | - | 4 - 5 | 345 | 97 |