Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HIF1AN
Entrez Gene ID
520864
Gene Name
hypoxia inducible factor 1 subunit alpha inhibitor
Gene Aliases
-
Chromosome Location
Chr.26: 21422732 - 21431815 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083443.2 NP_001076912.1 2 - 3 437 117
GenBank mRNA BC133607.1 - 2 - 3 438 117

View Full Product Details