Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM98A
Entrez Gene ID
530070
Gene Name
family with sequence similarity 98 member A
Gene Aliases
-
Chromosome Location
Chr.11: 16138053 - 16153872 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083455.1 NP_001076924.1 2 - 3 231 93
GenBank mRNA BC123587.1 - 2 - 3 231 93

View Full Product Details