Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIPK1A
Entrez Gene ID
541171
Gene Name
divergent protein kinase domain 1A
Gene Aliases
FAM69A
Chromosome Location
Chr.3: 50499771 - 50629763 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083500.1 NP_001076969.1 3 - 4 333 119
GenBank mRNA BC123747.1 - 3 - 4 333 119

View Full Product Details