Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CERS1
Entrez Gene ID
615296
Gene Name
ceramide synthase 1
Gene Aliases
LASS1
Chromosome Location
Chr.7: 4313437 - 4332887 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083514.2 NP_001076983.1 6 - 7 1024 96
GenBank mRNA BC114845.1 - 6 - 7 1025 96

View Full Product Details